Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD79B protein, rat (HEK293, His) | >95.0% | 10 UG
SDP

Catalog No. 5000290469
Encompass_Preferred

Recombinant rat CD79B protein fragment expressed in HEK293 cells with a C-terminal His tag, supplied as a purified sterile powder for use in biochemical and cell-based assays investigating B-cell receptor signaling.

  • Expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Partial sequence V26-D158 representing the extracellular fragment.
  • Purity >95% by reducing SDS-PAGE.
  • Approximate molecular weight 25-30 kDa (glycosylated).
  • Supplied sizes include 10 μg; additional sizes available on request.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.

Catalog No. 50-002-90469 Supplier Medchemexpress LLC Supplier No. HYP7565910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat CD79B protein fragment expressed in HEK293 cells with a C-terminal His tag, supplied as a purified sterile powder for use in biochemical and cell-based assays investigating B-cell receptor signaling.

  • Expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Partial sequence V26-D158 representing the extracellular fragment.
  • Purity >95% by reducing SDS-PAGE.
  • Approximate molecular weight 25-30 kDa (glycosylated).
  • Supplied sizes include 10 μg; additional sizes available on request.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.