Learn More
Sino Biological Recombinant Human CD79B Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 29750H08H100
Encompass_Preferred
A DNA sequence encoding the human CD79B (NP_000617.1) (Met1-Asp159) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. NC1556581