Learn More
ACROBiosystems Recombinant Protein;Human CD79B Protein, Fc Tag;HEK293;1MG;CDB,CD79b,B29,IGB,Ig-beta
Supplier:  ACROBiosystems CDBH52591MG
Encompass_Preferred
Recombinant Protein;Human CD79B Protein, Fc Tag;1MG;Human CD79B, Fc Tag (CDB-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # NP_000617).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.8 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8752