Learn More
Genscript Corporation CD8 alpha/CD8A His Human
Catalog No. 5000057287
Encompass
CD8 alpha enhances the responses of antigen-specific CTL activated through TCR through binding MHC class I favoring lipid raft partitioning of TCR and inducing intracellular signaling CD8 alpha is also found on dendritic cells and rat macrophages but whether CD8 alpha enhances responses of a partner receptor like TCR to activate these cells is not known
Catalog No. 50-000-57287 Supplier Genscript Corporation Supplier No. Z04228500
CD8 alpha enhances the responses of antigen-specific CTL activated through TCR through binding MHC class I favoring lipid raft partitioning of TCR and inducing intracellular signaling CD8 alpha is also found on dendritic cells and rat macrophages but whether CD8 alpha enhances responses of a partner receptor like TCR to activate these cells is not known