Learn More
Novus Biologicals™ CD8 Antibody Pair [HRP]

Description
Specifications
Specifications
| Assay Range | 78 pg/mL |
| Assay Sensitivity | 78-5000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Ferret CD8. |
| Target Species | Ferret |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD8, CD8a molecule, LEU2, p32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.