Learn More
CAPRICO BIOTECHNOLOGIES INC CD8 Antibody, 4109045
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4109045
Encompass_Preferred
The Caprico CD8 Antibody is a mouse monoclonal antibody that recognizes human CD8. It is produced in ascites and purified by protein A/G chromatography. This antibody is recommended for detection of CD8 alpha chain of human cytotoxic T lymphocytes by flow cytometry, immunohistochemistry on frozen sections, and Western blotting. The immunogen is human CD8 alpha chain extracellular domain. This antibody has been tested in flow cytometry, immunohistochemistry, and Western blot applications. The observed molecular weight of CD8 is 32-34 kDa. Each vial contains 200 μg IgG in 1.0 mL of PBS with < 0.1% sodium azide and 0.1% gelatin. This antibody is guaranteed for 6 months from date of receipt.
Catalog No. 50-227-9259