Learn More
Medchemexpress LLC CD8 beta Human HEK 100ug
SDP
Catalog No. 5000178374
Encompass_Preferred
CD8 beta is an important immune glycoprotein that serves as a coreceptor for MHC class I peptide complexes linking T cells to antigens presented by APCs It interacts with TCR and MHC class I recruits LCK kinase and initiates multiple signaling pathways CD8 beta Protein Human (HEK293) is the recombinant human-derived CD8 beta protein expressed by HEK293 with tag free
Catalog No. 50-001-78374 Supplier Medchemexpress LLC Supplier No. HYP74266100UG
CD8 beta is an important immune glycoprotein that serves as a coreceptor for MHC class I peptide complexes linking T cells to antigens presented by APCs It interacts with TCR and MHC class I recruits LCK kinase and initiates multiple signaling pathways CD8 beta Protein Human (HEK293) is the recombinant human-derived CD8 beta protein expressed by HEK293 with tag free