Learn More
Sino Biological Recombinant Human CD8 beta Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 11031HCCHB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD8B (P10966-1) (Met1-Pro170) was expressed with six amino acids (LEVLFQ) at the C- terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6234