Learn More
CAPRICO BIOTECHNOLOGIES INC CD8 mFlluor 450 Antibody, 4006145
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4006145
Encompass_Preferred
The Caprico CD8 MFluor 450 Antibody is a mouse monoclonal antibody that specifically binds to human CD8. It is conjugated to MFluor 450, a blue fluorescent dye with an excitation max of 450nm and emission max of 466nm. This antibody can be used for flow cytometry, immunofluorescence microscopy, and other fluorescent detection methods. It is recommended for staining of human peripheral blood cells, cell lines, and tissues. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100μl of a 0.2mg/ml antibody solution. The antibody should be stored at 4°C protected from prolonged exposure to light.
Catalog No. 50-227-9272