Learn More
CAPRICO BIOTECHNOLOGIES INC CD8 PerCP Antibody, 100 tests, 4109035
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4109035
Encompass_Preferred
The CD8 PerCP Antibody is a mouse monoclonal antibody that specifically binds to human CD8. It is conjugated to PerCP and is suitable for flow cytometry applications. This antibody recognizes the 32-34 kD α-chain of human CD8. Each vial contains 100 tests of antibody in PBS with 0.1% sodium azide and 0.2% BSA. The recommended dilution is 5-25 μl per test. This antibody is guaranteed for 6 months from date of receipt when stored at 2-8°C.
Catalog No. 50-227-9265