Learn More
Diagnostic Biosystems CD8 T-CELL CD8 (SP16)
Catalog No. 502407324
Encompass_Preferred
This antibody recognizes the CD8. The CD8 molecule consists of two chains, α and β, which are expressed as a disulphide-linked α/β heterodimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not β chain).
Catalog No. 50-240-7324 Supplier Diagnostic Biosystems Supplier No. RMPD012
This antibody recognizes the CD8. The CD8 molecule consists of two chains, α and β, which are expressed as a disulphide-linked α/β heterodimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not β chain).