Learn More
Enzo Life Sciences CD80 (human) monoclonal antibody (mAb 104) (Alexa Fluor 647) (100µg)
SDP
Catalog No. 502012712
Encompass_Preferred
Clone: mAb 104. Isotype: IgG1. Immunogen: Jijoye cells (human Burkitt’s lymphoma cell line). Species reactivity: Human. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2712 Supplier Enzo Life Sciences Supplier No. ENZABS1540100
Clone: mAb 104. Isotype: IgG1. Immunogen: Jijoye cells (human Burkitt’s lymphoma cell line). Species reactivity: Human. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.