Learn More
Enzo Life Sciences CD80 (human) monoclonal antibody (mAb 104) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS1560100
Encompass_Preferred
Clone: mAb 104. Isotype: IgG1. Immunogen: Jijoye cells (human Burkitt’s lymphoma cell line). Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In TRIS-NaCl, pH 8.
Catalog No. 50-201-2714