Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cd80-in-3 | 486449-65-4 | MFCD32701897 | 98.0% | 323.28 g/mol | C17H10FN3O3 | 100 MG
SDP

Catalog No. 5000212911
Encompass_Preferred

CD80-IN-3 is a small-molecule inhibitor that blocks the CD80/CD28 interaction, showing in vitro potency (EC50 = 630 nM) and measurable binding affinity (Kd = 125 nM). It is supplied as a solid and is also available as a DMSO solution for assay use; intended for research use only.

  • Potent CD80 inhibitor; inhibits CD80/CD28 interaction (EC50 = 630 nM).
  • Measured binding affinity Kd = 125 nM.
  • High purity solid suitable for biochemical and cellular assays (98.0%).
  • Molecular weight 323.28 g/mol; molecular formula C17H10FN3O3.
  • CAS number 486449-65-4 for unambiguous identification.
  • Available in milligram quantities and solution formats for assay flexibility.

Catalog No. 50-002-12911 Supplier Medchemexpress LLC Supplier No. HY100891100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CD80-IN-3 is a small-molecule inhibitor that blocks the CD80/CD28 interaction, showing in vitro potency (EC50 = 630 nM) and measurable binding affinity (Kd = 125 nM). It is supplied as a solid and is also available as a DMSO solution for assay use; intended for research use only.

  • Potent CD80 inhibitor; inhibits CD80/CD28 interaction (EC50 = 630 nM).
  • Measured binding affinity Kd = 125 nM.
  • High purity solid suitable for biochemical and cellular assays (98.0%).
  • Molecular weight 323.28 g/mol; molecular formula C17H10FN3O3.
  • CAS number 486449-65-4 for unambiguous identification.
  • Available in milligram quantities and solution formats for assay flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.