Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 975MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: 1.3.3.22. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Ramos, Daudi, Raji, U266, HUT-78, HUT-102, Jurkat, HL-60, KG1a, A293, A431, MG63, and human lymphocytes. Tonsil or Liver. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: B-Cell line derived from a Burkitt lymphoma. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 26kDa.Chromosome: 11p15.5. Gene Symbol: CD81. Gene ID: 975. Ensemble: ENSG00000110651. Uniprot: NP_001284578.
Catalog No. 50-188-5389