Learn More
Santa Cruz Biotechnology CD81 Antibody (5A6), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC23962
Encompass
CD81, also called TAPA-1, is a type III transmembrane protein that is broadly expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. CD81 is believed to be involved in both cell growth and signal transduction. It can be present as a multimolecular complex in association with CD37 and/or CD53, or on the surface of B cells in association with CD19, CD21 and/or MHC class II antigens.
Catalog No. NC1549099