Learn More
System Biosciences LLC CD81 EXO-FLOW CAPTURE KITCD81 Exo-Flow capture kit (Magnetic streptavidin beads, CD81-biotin capture antibody, Wash and Elution Buffers, Exo-FITC stain) = 10 Reactions
SDP
Catalog No. NC1913525
Encompass_Preferred
For the selective capture and flow sorting (FACS) of exosome subpopulations based on the presence of CD81, which is involved in cell targeting and adhesion. The CD81 Exo-Flow Capture Kit has all the reagents you need to purify exosomes using FACS—magnetic streptavidin beads, CD81-biotin capture antibody, wash and elution buffers, and reversible Exo-FITC stain (Cat.# EXOFLOW800A-1)—you supply the FACS and the optional magnetic stand (Cat.# EXOFLOW700A-1). Our well-validated CD81-biotin antibody and high-quality kit components ensure reliable, reproducible exosome purification based on the presence of CD81 on the exosome surface. And with our larger-than-typical bead size (9.1 μm diameter) exosome capture is highly efficient, assisting with recovery of rare exosome populations.
Catalog No. NC1913525 Supplier System Biosciences LLC Supplier No. EXOFLOW400A1
For the selective capture and flow sorting (FACS) of exosome subpopulations based on the presence of CD81, which is involved in cell targeting and adhesion. The CD81 Exo-Flow Capture Kit has all the reagents you need to purify exosomes using FACS—magnetic streptavidin beads, CD81-biotin capture antibody, wash and elution buffers, and reversible Exo-FITC stain (Cat.# EXOFLOW800A-1)—you supply the FACS and the optional magnetic stand (Cat.# EXOFLOW700A-1). Our well-validated CD81-biotin antibody and high-quality kit components ensure reliable, reproducible exosome purification based on the presence of CD81 on the exosome surface. And with our larger-than-typical bead size (9.1 μm diameter) exosome capture is highly efficient, assisting with recovery of rare exosome populations.