Learn More
Medchemexpress LLC CD81 Human HEK293 50ug
SDP
Catalog No. 5000178560
Encompass_Preferred
The CD19 receptor is critical for B cell activation driving the trafficking and assembly of CD19-CR2 and BCR complexes thereby lowering the antigenic threshold for B cell clonal expansion In T cells CD19 contributes to CD3 localization and influences polarization CD81 Protein Human (HEK293 His) is the recombinant human-derived CD81 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-78560 Supplier Medchemexpress LLC Supplier No. HYP7426450UG
The CD19 receptor is critical for B cell activation driving the trafficking and assembly of CD19-CR2 and BCR complexes thereby lowering the antigenic threshold for B cell clonal expansion In T cells CD19 contributes to CD3 localization and influences polarization CD81 Protein Human (HEK293 His) is the recombinant human-derived CD81 protein expressed by HEK293 with N-His labeled tag