Learn More
CAPRICO BIOTECHNOLOGIES INC CD81 (MF450), 4056145
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4056145
Encompass_Preferred
CD81 (MF450) is a mouse monoclonal antibody from Caprico Biotechnologies. It is produced in ascites and recognizes human CD81. This antibody can be used for ELISA, Western Blot, Immunohistochemistry, and Flow Cytometry applications. The immunogen is human CD81 extracellular domain. This antibody is purified by protein A/G.
Catalog No. 50-227-9277