Learn More
Sino Biological CD81 / TAPA-1 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 14244MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD81 / TAPA-1 (rh CD81 / TAPA-1; Catalog#14244-HNCH; NP_004347.1; Phe113-Lys201). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8418