Learn More
Enzo Life Sciences CD83 (human) monoclonal antibody (1G11) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX805045C050
Encompass_Preferred
Clone: 1G11. Isotype: IgG1. Immunogen: Recombinant human CD83 (extracellular domain). Species reactivity: Human. Specificity: The binding affinity strongly increases after deglycosylation of mature DC. Applications: Flow Cytometry, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-0042