Learn More
ACROBiosystems Recombinant Protein;Human CD83 Protein;HEK293;100UG;CD3,CD83,HB15,hCD83,BL11,HB-15
Catalog No. 501658070
Encompass_Preferred
Recombinant Protein;Human CD83 Protein;100UG;Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Ala 143 (Accession # AAH30830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 18 kDa and 22 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-8070 Supplier ACROBiosystems Supplier No. CD3H5223100UG
Recombinant Protein;Human CD83 Protein;100UG;Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Ala 143 (Accession # AAH30830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 18 kDa and 22 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.