Learn More
Aviva Systems Biology Corp CD84 Antibody - C-terminal region (ARP64101_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64101_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD84. It was validated on Western Blot. . Alternate names: DKFZp781E2378, LY9B, SLAMF5, hCD84, mCD84ApplicationWB. Species reactivity: Human
Catalog No. 50-288-174