Learn More
Biolegend APC anti-human CD84
Supplier:  Biolegend 326010
Encompass_Preferred
APC anti-human CD84 [CD84.1.21]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1922722