Learn More
ACROBiosystems Recombinant Protein;Human CD84 / SLAMF5 Protein;HEK293;1MG;CD4,CD84,SLAMF5,LY9B,SLAMF5,hCD84,mCD84,SLAMF-5
Supplier:  ACROBiosystems CD4H52291MG
Encompass_Preferred
Recombinant Protein;Human CD84 / SLAMF5 Protein;1MG;Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession # AAH20063).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8303