Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD84 / SLAMF5 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;CD4,CD84,SLAMF5,LY9B,SLAMF5,hCD84,mCD84,SLAMF-5
Catalog No. 501657702
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD84 / SLAMF5 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CD84, His Tag, Avi Tag (CD4-H82E5) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession # Q9UIB8-3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 40-50 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7702 Supplier ACROBiosystems Supplier No. CD4H82E525UG
Recombinant Protein;Biotinylated Human CD84 / SLAMF5 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CD84, His Tag, Avi Tag (CD4-H82E5) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession # Q9UIB8-3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 40-50 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.