Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 942MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM600. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: 293T or Jukat cells. Monocytes, dendritic cells, activated T, B, natural killer cells in lymph node or tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length CD86 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 70kDa.Chromosome: 3q13.33. Gene Symbol: CD86. Gene ID: 942. Ensemble: ENSG00000114013. Uniprot: NP_787058.
Catalog No. 50-188-5339