Learn More
Sino Biological CD86 Antibody (APC), Mouse MAb 25Tests
Supplier:  Sino Biological 80025MM10A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD86 (rR CD86; Catalog#80025-R08H; NP_064466.1; Met1-Ile250) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-6551