Learn More
Beckman Coulter Beckman Coulter CD86-PE antibody, clone HA5.2B7 (IM2729U)
Supplier:  Beckman Coulter IM2729U
Encompass_Preferred
Beckman Coulter CD86-PE (IM2729U) is a PE-conjugated monoclonal antibody (clone HA5.2B7) designed for flow cytometry detection of CD86 (B7-2) on human immune cells. Supplied as a ready-to-use conjugate, it supports immunophenotyping and activation-state analysis.
  • PE-conjugated monoclonal antibody suitable for flow cytometry.
  • Clone HA5.2B7 provides specific binding to CD86.
  • Mouse-derived antibody with IgG2b isotype.
  • Ready-to-use conjugate supplied in a 2 mL format.
  • Validated for detection of CD86 on human leukocytes.
  • Useful for immunophenotyping and activation studies.
Catalog No. NC0546754