Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 925MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C8/144B. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HuT78 or hPBL. Tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 32kDa.Chromosome: 2p11.2. Gene Symbol: CD8A. Gene ID: 925. Ensemble: ENSG00000153563. Uniprot: NP_741969.
Catalog No. 50-188-5329