Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 925RBM9P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: C8/1779R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG kappa. Positive Control: HuT78 or hPBL. Tonsil. Cellular Staining: Cell Surface. Buffer: 10mM PBS with or without 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human CD8a protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 32kDa.Chromosome: 2p11.2. Gene Symbol: CD8A. Gene ID: 925. Ensemble: ENSG00000153563. Uniprot: NP_741969.
Catalog No. 50-188-5331