Learn More
Aviva Systems Biology Corp CD8A Antibody - middle region (ARP59114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD8A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CD8A. Alternate names: CD8, Leu2, MAL, p32ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-286-584