Learn More
Enzo Life Sciences CD8 alpha Recombinant monoclonal antibody (YTS 105.18) (Rat IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4570200
Encompass_Preferred
Clone: YTS 105.18. Isotype: IgG2aκ. Immunogen: Ly-2-transfected mouse L cells. Species reactivity: Mouse. Specificity: Recognizes a non polymorphic epitope on the mouse CD8 α chain. Applications: Flow Cytometry, IF, IHC (FS), FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2984