Learn More
Aviva Systems Biology Corp CD8B Antibody - middle region (ARP42205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD8B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CD8B. Alternate names: CD8B1, LYT3, Leu2, Ly3, MGC119115, LY3, P37, LEU2ApplicationWB. Species reactivity: Dog, Human, Rabbit, Rat
Catalog No. 50-284-983