Learn More
Aviva Systems Biology Corp CD8B Antibody - N-terminal region (ARP42204_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42204_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD8B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CD8B. Alternate names: CD8B, LYT3, Leu2, Ly3, LY3, P37, LEU2, CD8B1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-54060