Learn More
Enzo Life Sciences CD8 beta Recombinant monoclonal antibody (YTS 156.7.7) (Rat IgG2b kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6310200
Encompass_Preferred
Clone: YTS 156.7.7. Isotype: IgG2bκ. Immunogen: Mouse CD8 (lyt-3). Species reactivity: Mouse. Specificity: The antibody recognizes the B epitope of the mouse Lyt-3 (CD8) antigen. Has been used in CD8 subset depletion in in vivo and flow cytometry studies. Applications: Flow Cytometry, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3155