Learn More
Sino Biological Anti-CD8 alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10980T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD8/CD8 alpha/Leu-2 (rh CD8/CD8 alpha/Leu-2; Catalog#10980-H08H; NP_001759.3; Met1-Asp182). CD8/CD8 alpha/Leu-2 specific IgG was purified by Human CD8/CD8 alpha/Leu-2 affinity chromatography.
Catalog No. 50-222-3373