Learn More
STEMCELL Technologies Anti-Human CD9 Antibody, Clone HI9a, Size: 100 ug
SDP
Supplier:  STEMCELL Technologies 1000138
Encompass
The HI9a antibody reacts with an extracellular epitope on human CD9 a 24 kDa single-chain type III transmembrane glycoprotein. CD9 is broadly expressed including on the surface of monocytes developing B cells activated T cells granulocytes epithelial cells endothelial cells and platelets (mainly the a-granules) and is also a marker characteristic of extracellular vesicles.
Catalog No. 50-239-0277