Learn More
System Biosciences LLC Anti-CD9 Antibody with Goat Anti-Rabbit HRP Secondary Antibody, 25 uL
SDP
Supplier:  System Biosciences LLC EXOABCD9A1
Encompass
Detect exosomes using SBI’s rigorously validated polyclonal rabbit anti-CD9 antibodies that recognize the human and mouse versions of the protein.
Catalog No. NC0724360