Learn More
Aviva Systems Biology Corp CD9 Antibody - N-terminal region (ARP61171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CD9. Alternate names: 5H9, BA2, BTCC-1, DRAP-27, GIG2, MIC3, MRP-1, P24, TSPAN29, TSPAN-29ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-281