Learn More
Enzo Life Sciences CD9 (human) monoclonal antibody (Gi15) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805027C100
Encompass_Preferred
Clone: Gi15. Isotype: IgG2b. Immunogen: Human platelets. Species reactivity: Human. Applications: IP, WB. Formulation: Liquid. 100µg of purified antibody in 200µl PBS containing 0.05% sodium azide.
Catalog No. 50-201-0038