Learn More
Medchemexpress LLC CD9 Human HEK293 5ug
SDP
Catalog No. 5000182375
Encompass_Preferred
CD9 protein is an integral membrane protein that plays a critical regulatory role in processes such as sperm-egg fusion platelet activation and cell adhesion CD9 is critical for sperm-egg fusion coordinating multi-protein complexes on the oocyte surface CD9 Protein Human (HEK293 His) is the recombinant human-derived CD9 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82375 Supplier Medchemexpress LLC Supplier No. HYP727015UG
CD9 protein is an integral membrane protein that plays a critical regulatory role in processes such as sperm-egg fusion platelet activation and cell adhesion CD9 is critical for sperm-egg fusion coordinating multi-protein complexes on the oocyte surface CD9 Protein Human (HEK293 His) is the recombinant human-derived CD9 protein expressed by HEK293 with C-6 His labeled tag