Learn More
Genscript Corporation CD93/C1q R1 hFc Chimera Human
Supplier:  Genscript Corporation Z04233500
Encompass
CD93 has been shown critical roles in inflammatory and immune diseases CD93 silencing increased IL-6 and TSLP but not IL-33 levels in culture supernatants HDM-induced asthma mice showed significant airway hyperresponsiveness and inflammation with Th2 cytokine activation along with decreased CD93 expression in bronchial epithelial cells and lung homogenates but increased serum CD93 levels
Catalog No. 50-000-56941