Learn More
Sino Biological CD93 / C1qR Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 12589MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD93 / C1qR (rh CD93 / C1qR; Catalog#12589-H08H; Q9NPY3; Met1-Lys580). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7437