Learn More
Genscript Corporation CD94 His Avi Human
Catalog No. 5000057691
Encompass
CD94 is an approximately 25 kDa type 2 transmembrane protein that plays an important role in regulating natural killer (NK) cell activation CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells
Catalog No. 50-000-57691 Supplier Genscript Corporation Supplier No. Z04235500
CD94 is an approximately 25 kDa type 2 transmembrane protein that plays an important role in regulating natural killer (NK) cell activation CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells