Learn More
ACROBiosystems ACROBiosystems Human CD94 Protein, His Tag
Catalog No. 501956801
Encompass_Preferred
Recombinant Protein;100UG;Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.1 kDa. The protein migrates as 24-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6801 Supplier ACROBiosystems Supplier No. CD4H52H8100UG
Recombinant Protein;100UG;Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.1 kDa. The protein migrates as 24-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.