Learn More
Medchemexpress LLC CD94 Human HEK293 50ug
SDP
Catalog No. 5000182023
Encompass_Preferred
CD94 protein is a key immune receptor for self-non-self discrimination It forms a complex with KLRC1 or KLRC2 on lymphocyte subsets and recognizes HLA-E loaded with self-peptides It allows cytotoxic cells to monitor MHC class I expression and promote self-tolerance CD94 Protein Human (HEK293 His) is the recombinant human-derived CD94 protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-82023 Supplier Medchemexpress LLC Supplier No. HYP7270050UG
CD94 protein is a key immune receptor for self-non-self discrimination It forms a complex with KLRC1 or KLRC2 on lymphocyte subsets and recognizes HLA-E loaded with self-peptides It allows cytotoxic cells to monitor MHC class I expression and promote self-tolerance CD94 Protein Human (HEK293 His) is the recombinant human-derived CD94 protein expressed by HEK293 with N-6 His labeled tag