Learn More
Enzo Life Sciences CD94 monoclonal antibody (AT13E3) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7230100
Encompass_Preferred
Clone: AT13E3. Isotype: IgG1κ. Immunogen: Recombinant human KLRD1 (aa 32-179) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3264