Learn More
ACROBiosystems ACROBiosystems Human CD94 Protein, Mouse IgG2a Fc Tag
Catalog No. 501956877
Encompass_Preferred
Recombinant Protein;1MG;Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6877 Supplier ACROBiosystems Supplier No. CD4H52531MG
Recombinant Protein;1MG;Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.