Learn More
Genscript Corporation CD96/TACTILE His Mouse
Supplier:  Genscript Corporation Z04333100
Encompass
The receptors CD96 and TIGIT are expressed on the surface of T and natural killer (NK) cells and recent studies suggest both play important inhibitory roles in immune function CD96 has been shown to modulate immune cell activity in mice with Cd96-/- mice displaying hypersensitive NK-cell responses to immune challenge and significant tumor resistance The counterbalance between the putative inhibitory CD96 and TIGIT receptors and the activating receptor CD226 offers unique strategies for immuno-oncology drug development
Catalog No. 50-000-59136